Lithium acetoacetate ≥97.0%
Supplier: TCI America
Synonyms:
3-Oxobutyric acid Lithium salt, Lithium 3-Oxobutyrate, Acetoacetic acid Lithium salt
A1478-5G
A1478-1G
TCA1478-5GEA
365.18
USD
TCA1478-5G
TCA1478-001G
Lithium acetoacetate ≥97.0%
Lithium acetoacetate
CAS Number: 3483-11-2
MDL Number: MFCD00013060
Molecular Formula: C4H6O3
Molecular Weight: 108.02
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Storage Temperature: <0°C
Other Product Names / Synonyms: Acetoacetic Acid Lithium Salt, Lithium 3-Oxobutyrate, 3-Oxobutyric Acid Lithium Salt
Formula:
C₄H₅LiO₃ |
MDL Number:
MFCD00013060 CAS Number: 3483-11-2 |
|
Specification Test Results
Purity/Analysis Method | >97.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...