4-Aminobenzylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 4-(Aminomethyl)aniline, alpha-Amino-p-toluidine

A1436-25G A1436-250G
TCA1436-025GEA 173 USD
TCA1436-025G TCA1436-250G
4-Aminobenzylamine ≥98.0% (by GC, titration analysis)
4-Aminobenzylamine

CAS Number: 4403-71-8
MDL Number: MFCD00075513
Molecular Formula: C7H10N2
Molecular Weight: 122.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Flash Point (°C): 113
Freezing point (°C): 24
Lambda max.: 246 nm (EtOH)


Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials
Other Product Names / Synonyms: 4-(Aminomethyl)aniline, alpha-Amino-p-toluidine

Formula: C₇H₁₀N₂
MW: 122.17 g/mol
Flash Pt: 113 °C
MDL Number: MFCD00075513
CAS Number: 4403-71-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=64ec6d792672e58cd620cf8a5305bdeddd5e2d43&vid=64ec6d792672e58cd620cf8a5305bdeddd5e2d43&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR