3-Aminobenzylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: alpha-Amino-m-toluidine, 3-(Aminomethyl)aniline

A1431-25G A1431-5G
TCA1431-025GEA 126.6 USD
TCA1431-025G TCA1431-005G
3-Aminobenzylamine ≥98.0% (by GC, titration analysis)
3-Aminobenzylamine

CAS Number: 4403-70-7
MDL Number: MFCD00078355
Molecular Formula: C7H10N2
Molecular Weight: 122.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 134
Melting point (°C): 43


Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials
Other Product Names / Synonyms: 3-(Aminomethyl)aniline, alpha-Amino-m-toluidine

Formula: C₇H₁₀N₂
MW: 122.17 g/mol
Boiling Pt: 134 °C
Melting Pt: 43 °C
MDL Number: MFCD00078355
CAS Number: 4403-70-7
EINECS: 412-230-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e7b7d3ffd56fbf1f981dae37285e756bbf2cbf59&vid=e7b7d3ffd56fbf1f981dae37285e756bbf2cbf59&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR