6-Amino-1,3-dimethyluracil ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
6-Amino-1,3-dimethylpyrimidine-2,4(1H,3H)-dione, 4-Amino-1,3-Dimethyluracil
A1420-25G
A1420-500G
TCA1420-025GEA
35
USD
TCA1420-025G
TCA1420-500G
6-Amino-1,3-dimethyluracil ≥98.0% (by HPLC, titration analysis)
6-Amino-1,3-dimethyluracil
CAS Number: 6642-31-5
MDL Number: MFCD00006552
Molecular Formula: C6H9N3O2
Molecular Weight: 155.16
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Formula:
C₆H₉N₃O₂ MW: 155.1546 g/mol |
MDL Number:
MFCD00006552 CAS Number: 6642-31-5 EINECS: 229-662-0 |
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...