2-Aminoindan ≥99.0% (by GC, titration analysis)

Supplier: TCI America
Warning

A1320-25G A1320-5G
TCA1320-25GEA 317 USD
TCA1320-25G TCA1320-5G
2-Aminoindan ≥99.0% (by GC, titration analysis)
2-Aminoindan

CAS Number: 2975-41-9
MDL Number: MFCD00082597
Molecular Formula: C9H11N
Molecular Weight: 133.19
Purity/Analysis Method: >99.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 229
Specific Gravity (20/20): 1.03


Other Product Names / Synonyms: 2-Indanamine

Formula: C₉H₁₁N
Boiling Pt: 229 °C
Density: 1.03 (20 °C)
MDL Number: MFCD00082597
CAS Number: 2975-41-9
EINECS: 221-021-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1f3ab88e25acec7cea5a000a3904eec1de600fe3&vid=1f3ab88e25acec7cea5a000a3904eec1de600fe3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR