4-(2-Aminoethyl)pyridine ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2-(4-Pyridyl)ethylamine, 2-Pyridin-4-yl-ethylamine

A1264-5G A1264-25G
TCA1264-005GEA 145.65 USD
TCA1264-005G TCA1264-025G
4-(2-Aminoethyl)pyridine ≥97.0% (by titrimetric analysis)
4-(2-Aminoethyl)pyridine

CAS Number: 13258-63-4
MDL Number: MFCD00038045
Molecular Formula: C7H10N2
Molecular Weight: 122.17
Purity/Analysis Method: >97.0% (T)
Form: Clear Liquid
Boiling point (°C): 120
Flash Point (°C): 110
Specific Gravity (20/20): 1.03


Other Product Names / Synonyms: 2-(4-Pyridyl)ethylamine

Formula: C₇H₁₀N₂
MW: 122.17 g/mol
Boiling Pt: 120 °C
Melting Pt: 76 °C
Density: 1.03 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00038045
CAS Number: 13258-63-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=62d8fca73320205695849b7b075cc2437c1a33ea&vid=62d8fca73320205695849b7b075cc2437c1a33ea&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR