2-(Aminomethyl)-1,3-dioxolane ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A1179-5G A1179-1G
TCA1179-5GEA 502.6 USD
TCA1179-5G TCA1179-001G
2-(Aminomethyl)-1,3-dioxolane ≥98.0% (by GC, titration analysis)
2-(Aminomethyl)-1,3-dioxolane

CAS Number: 4388-97-0
MDL Number: MFCD00142862
Molecular Formula: C4H9NO2
Molecular Weight: 103.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Specific Gravity (20/20): 1.10

Formula: C₄H₉NO₂
MW: 103.12 g/mol
Density: 1.10 (20 °C)
MDL Number: MFCD00142862
CAS Number: 4388-97-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b74dd59c804ff0872de8008310a89f8a3dd19732&vid=b74dd59c804ff0872de8008310a89f8a3dd19732&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR