1-Aminocyclopropanecarboxylic acid ≥97.0% (by titrimetric analysis)

Supplier: TCI America

A1178-100MG A1178-1G A1178-5G
TCA1178-100MGEA 18.32 USD
TCA1178-100MG TCA1178-1G 76218-730
1-Aminocyclopropanecarboxylic acid ≥97.0% (by titrimetric analysis)
1-Aminocyclopropanecarboxylic acid

CAS Number: 22059-21-8
MDL Number: MFCD00009944
Molecular Formula: C4H7NO2
Molecular Weight: 101.11
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 231


Product Categories: Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: Carboxycyclopropylamine

Formula: C₄H₇NO₂
MW: 101.11 g/mol
Melting Pt: 231 °C
MDL Number: MFCD00009944
CAS Number: 22059-21-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e314973971dd439e489b0bf22e3b09ac41a10ec2&vid=e314973971dd439e489b0bf22e3b09ac41a10ec2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR