Aminoguanidine hydrochloride ≥98.0%
Supplier: TCI America
Synonyms:
Aminoguanidine HCl, Aminoguanidine Hydrochloride, Guanylhydrazine hydrochloride
A1129-25G
A1129-500G
A1129-100G
TCA1129-025GEA
55.95
USD
TCA1129-025G
TCA1129-500G
TCA1129-100G
Aminoguanidine hydrochloride ≥98.0%
Aminoguanidine hydrochloride
CAS Number: 1937-19-5
MDL Number: MFCD00039074
Molecular Formula: CH6N4
Molecular Weight: 110.55
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 165
Formula:
CH₇ClN₄ MW: 110.55 g/mol Melting Pt: 165 °C |
MDL Number:
MFCD00039074 CAS Number: 1937-19-5 EINECS: 217-707-7 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...