(S)-(+)-2-Amino-1-propanol ≥98.0%

Supplier: TCI America
Danger

Synonyms: L(+)-Alaninol, L-Alaninol, (S)-2-Aminopropan-1-ol, L(+)-2-Amino-1-propanol

A1085-5ML A1085-1ML A1085-25ML
TCA1085-005MLEA 94 USD
TCA1085-005ML TCA1085-001ML TCA1085-25ML
(S)-(+)-2-Amino-1-propanol ≥98.0%
(S)-(+)-2-Amino-1-propanol

CAS Number: 2749-11-3
MDL Number: MFCD00064412
Molecular Formula: C3H9NO
Molecular Weight: 75.11
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 98
Flash Point (°C): 83
Specific Gravity (20/20): 0.97
Specific rotation [a]20/D: 22 deg (C=2, EtOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: L-Alaninol

Formula: C₃H₉NO
Boiling Pt: 98 °C
Density: 0.97 (20 °C)
Flash Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00064412
CAS Number: 2749-11-3
EINECS: 220-388-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8e48c8c68aa05e3f0bf5082d65fb35306a765752&vid=8e48c8c68aa05e3f0bf5082d65fb35306a765752&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR