Heptyl acetate ≥99.0%

Supplier: TCI America

Synonyms: Acetic acid heptyl ester

A0904-500ML A0904-25ML
TCA0904-500MLEA 215.72 USD
TCA0904-500ML TCA0904-025ML
Heptyl acetate ≥99.0%
Heptyl acetate

CAS Number: 112-06-1
MDL Number: MFCD00027311
Molecular Formula: C9H18O2
Molecular Weight: 158.24
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 192
Melting point (°C): -50
Flash Point (°C): 69
Specific Gravity (20/20): 0.87


Other Product Names / Synonyms: Acetic Acid Heptyl Ester

Formula: C₉H₁₈O₂
MW: 158.24 g/mol
Boiling Pt: 192 °C
Melting Pt: –50 °C
Density: 0.87 (20 °C)
Flash Pt: 69 °C
MDL Number: MFCD00027311
CAS Number: 112-06-1
EINECS: 203-932-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1dcf28335ca6c00a59a3e518a48966c90121e2ba&vid=30e3c0b89a636833cfe6f37993f933c3a7b36f3a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR