2-Amino-5-methyl-1,3,4-thiadiazole ≥98.0% (by titrimetric analysis)

Supplier: TCI America

A0874-25G A0874-5G
TCA0874-025GEA 176.8 USD
TCA0874-025G TCA0874-5G
2-Amino-5-methyl-1,3,4-thiadiazole ≥98.0% (by titrimetric analysis)
2-Amino-5-methyl-1,3,4-thiadiazole

CAS Number: 108-33-8
MDL Number: MFCD00003110
Molecular Formula: C3H5N3S
Molecular Weight: 115.15
Purity/Analysis Method: >98.0% (T)
Form: Crystal

Formula: C₃H₅N₃S
MW: 115.16 g/mol
MDL Number: MFCD00003110
CAS Number: 108-33-8
EINECS: 203-573-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ffba6e62ed8aa30f52ba47ab0c8f2dcec092fb7&vid=6ffba6e62ed8aa30f52ba47ab0c8f2dcec092fb7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR