5-Amino-2-naphthol ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

A0359-25G A0359-5G
TCA0359-025GEA 2227 USD
TCA0359-025G TCA0359-005G
5-Amino-2-naphthol ≥97.0% (by HPLC, titration analysis)
5-Amino-2-naphthol

CAS Number: 86-97-5
MDL Number: MFCD00035729
Molecular Formula: C10H9NO
Molecular Weight: 159.19
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Melting point (°C): 183

Formula: C₁₀H₉NO
MW: 159.19 g/mol
Melting Pt: 183 °C
MDL Number: MFCD00035729
CAS Number: 86-97-5
EINECS: 201-713-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d10531b218e35937ebffc5a4ab6a081308d19365&vid=d10531b218e35937ebffc5a4ab6a081308d19365&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR