Xylenol orange indicator 0,2% (w/v) in alcohol indicator grade
Supplier: Ricca Chemical
|
Danger
|
R9388000-500A
R9388000-120A
RC9388-16EA
727
USD
RC9388-16
RC9388-4
Xylenol orange indicator 0,2% (w/v) in alcohol indicator grade
Xylenol orange solutions
Formula:
C₃₁H₂₈N₂Na₄O₁₃S |
MDL Number:
MFCD00044293 CAS Number: 3618-43-7 UN: 1987 ADR: 3,II |
Specification Test Results
Appearance | Clear, red, alcohol odor |
Suitability for Use | Purple (Excess Metal) - Yellow (Excess EDTA) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...