Thymolphthalein 0.1% (w/v) in alcohol

Supplier: Ricca Chemical
Danger

Synonyms: 3,3-Bis(4-hydroxy-5-isopropyl-o-tolyl)phthalide, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)isobenzofuran-1(3H)-one

R8460000-500A R8460000-120A
RC8460-16EA 156 USD
RC8460-16 RC8460-4
Thymolphthalein 0.1% (w/v) in alcohol
Thymolphthalein
Formula: C₂₈H₃₀O₄
MW: 430.54 g/mol
Boiling Pt: ~ 77 °C
Density: 0.8000 g/cm³
Flash Pt: ~ 15.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00005909
CAS Number: 125-20-2
UN: 1987
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear, colorless, alcohol odor
Suitability for Use Colorless (pH 10.5)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR