Thioacetamide 4% (w/v) in aqueous solution USP Test Solution (TS)

Supplier: Ricca Chemical
Danger

Synonyms: Ethanethioamide

R8393400-500C R8393400-120C
RC8393.40-16EA 110 USD
RC8393.40-16 RC8393.40-4
Thioacetamide 4% (w/v) in aqueous solution USP Test Solution (TS)
Thioacetamide
Formula: CH₃CSNH₂
MW: 75.13 g/mol
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0200 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00008070
CAS Number: 62-55-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe2000e62db991c85340c257ac6d64ca42c3551a&vid=18e76b2cdee9ee22a2618ec371858a498f218457&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear, colorless, odorless to slight mercaptan odor
Refractive Index Confirmed Concentration

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR