Disodium-L(+)-tartrate 10% (w/v) in aqueous solution

Supplier: Ricca Chemical

R7850000-1A R7850000-500A
RC7850-32EA 112 USD
RC7850-32 RC7850-16
Disodium-L(+)-tartrate 10% (w/v) in aqueous solution
Disodium-L(+)-tartrate solutions
Formula: NaOOC(CHOH)₂COONa·2H₂O
Boiling Pt: ~ 100 °C
Density: 1.0700 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00150035
CAS Number: 6106-24-7

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Refractive Index Confirmed Concentration


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR