Sodium phenoxide solution for ammonia nitrogen determination

Supplier: Ricca Chemical
Danger

Synonyms: Sodium phenylate

R7518000-500A R7518000-1A
RC7518-16EA 103 USD
RC7518-16 RC7518-32
Sodium phenoxide solution for ammonia nitrogen determination
Sodium phenoxide
Formula: C₆H₅NaO
MW: 116.09 g/mol
Boiling Pt: ~ 100 °C
Density: 1.1000 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00013134
CAS Number: 139-02-6
UN: 3267
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear, tan to reddish, phenol odor
Suitability for Use Passes Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR