Sodium hydroxide 0.1142 N

Supplier: Ricca Chemical

R7355000-4A R7355000-20F
RC73551EA 94.52 USD
RC73551 RC73555
Sodium hydroxide 0.1142 N
Sodium hydroxide
Formula: NaOH
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1823
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 84) 0.1142 ± 0.0001 N at 20.0 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR