tri-Sodium citrate 30% (w/v) in aqueous solution

Supplier: Ricca Chemical

R7240000-500A R7240000-1A
RC724016EA 265 USD
RC724016 RC7240-32
tri-Sodium citrate 30% (w/v) in aqueous solution
tri-Sodium citrate solutions
Formula: Na₃C₆H₅O₇·2H₂O
MW: 294.1 g/mol
Boiling Pt: ~ 100 °C
Density: 1.2000 g/cm³
MDL Number: MFCD00150031
CAS Number: 6132-04-3

Order Now

Specification Test Results

Appearance Clear, colorless, odorless

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR