di-Ammonium oxalate 4% (w/v) in aqueous solution
Supplier: Ricca Chemical
|
Warning
|
R0685000-500A
R0685000-1A
RC685-16EA
74
USD
RC685-16
RC685-32
di-Ammonium oxalate 4% (w/v) in aqueous solution
di-Ammonium oxalate solutions
Formula:
(NH₄)₂C₂O₄·H₂O MW: 142.11 g/mol Boiling Pt: ~ 100 °C Melting Pt: ~ 0 °C Density: 1.0400 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00149694 CAS Number: 6009-70-7 |
Specification Test Results
Appearance | Clear, colorless, odorless |
Assay at 20 °C (traceable to NIST SRM 40) | 4.00 ± 0.01 % (w/v) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...