Ammonium heptamolybdate 4% (w/v) in aqueous solution

Supplier: Ricca Chemical
Warning

R0665000-500A R0665000-1A R0665000-4A
RC66516EA 126 USD
RC66516 RC665-32 RC665-1
Ammonium heptamolybdate 4% (w/v) in aqueous solution
Ammonium heptamolybdate solutions
Formula: (NH₄)₆Mo₇O₂₄·4H₂O
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0400 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00167059
CAS Number: 12054-85-2

Order Now

Specification Test Results

Appearance Clear, colorless


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR