Potassium iodate-iodide 0.100 N (N/10)

Supplier: Ricca Chemical

R6285000-1C R6285000-20F R6285000-4C
RC6285-32EA 173 USD
RC6285-32 RC6285-5 RC6285-1
Potassium iodate-iodide 0.100 N (N/10)
Potassium iodate-iodide

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 136) 0.1000 ± 0.0001 N at 20.0 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR