Potassium hexacyanoferrate(II) 10% (w/v) in aqueous solution
Supplier: Ricca Chemical
Synonyms:
Potassium ferrocyanide, Tetrapotassium hexacyanoferrate
R6096000-1A
R6096000-500A
RC6096-32EA
193
USD
RC6096-32
RC6096-16
Potassium hexacyanoferrate(II) 10% (w/v) in aqueous solution
Potassium hexacyanoferrate(II)
Formula:
C₆FeK₄N₆ MW: 368.35 g/mol Boiling Pt: ~ 100 °C Density: 1.0530 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00167022 CAS Number: 13943-58-3 |
Specification Test Results
Appearance | Clear, yellow, odorless |
Assay | 10.0 ± 0.3 % (w/v) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...