Potassium dichromate 0.500 N (N/2)

Supplier: Ricca Chemical

R6074000-1A R6074000-500A
RC6074-32EA 106 USD
RC6074-32 RC6074-16
Potassium dichromate 0.500 N (N/2)
Potassium dichromate
Formula: K₂Cr₂O₇
MW: 294.18 g/mol
Boiling Pt: 500
Melting Pt: 398
MDL Number: MFCD00011367
CAS Number: 7778-50-9
UN: 3087
ADR: 5.1 II
Merck Index: 12,07790

Order Now

Specification Test Results

Appearance Clear, orange, odorless
Assay at 20 °C (traceable to NIST SRM 136) 0.5000 ± 0.0001 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR