Potassium hydrogen diiodate 0.00235 M (0.0282 N)
Supplier: Ricca Chemical
Synonyms:
Potassium bi-iodate
R5877000-1C
R5877000-4C
RC587732EA
179
USD
RC587732
RC5877-1
Potassium hydrogen diiodate 0.00235 M (0.0282 N)
Potassium hydrogen diiodate
1 mL = 1 mg Cl2 1000 ppm
Formula:
HI₂KO₆ MW: 389.91 |
MDL Number:
MFCD00011400 CAS Number: 13455-24-8 UN: 1479 ADR: 5.1,II |
Specification Test Results
Appearance | Clear, colorless, odorless |
Assay at 20 °C (traceable to NIST SRM 136) | 0.02820 ± 0.00001 N at 20.0 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...