Phenylarsine oxide 0.0250 N (N/40) stabilised titrant

Supplier: Ricca Chemical

Synonyms: PAO, Oxophenylarsine, Arzene

R5755000-1C R5755000-4C
RC575532EA 515 USD
RC575532 RC5755-1
Phenylarsine oxide 0.0250 N (N/40) stabilised titrant
Phenylarsine oxide
Formula: C₆H₅AsO
MDL Number: MFCD00001990
CAS Number: 637-03-6
UN: 3465
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear to hazy, colorless to light tan, odorless
Assay at 20 °C (traceable to NIST SRM 136) 0.02500 ± 0.00002 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR