Phenolphthalein 1% (w/v) in pyridine indicator
Supplier: Ricca Chemical
|
Danger
|
Synonyms:
3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one
R5627000-500C
R5627000-1C
RC5627-16EA
125
USD
RC5627-16
RC5627-32
Phenolphthalein 1% (w/v) in pyridine indicator
Phenolphthalein
Formula:
C₂₀H₁₄O₄ Boiling Pt: 115…116 °C Melting Pt: –42 °C Density: 0.9830 g/cm³ Flash Pt: 20 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005913 CAS Number: 77-09-8 UN: 1282 ADR: 3,II |
|
Specification Test Results
Appearance | Clear, light yellow-green, pungent pyridine odor |
Suitability for Use | Colorless (pH 8.0) - Pink (pH 10.0) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...