Phenolphthalein 0.08% (w/v) in methanol indicator
Supplier: Ricca Chemical
|
Danger
|
Synonyms:
3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one
R5595000-120A
R5595000-500A
RC5595-4EA
47
USD
RC5595-4
RC5595-16
Phenolphthalein 0.08% (w/v) in methanol indicator
Phenolphthalein
Formula:
C₂₀H₁₄O₄ MW: 318.33 g/mol Boiling Pt: ~ 64.5 °C Melting Pt: –98 °C Density: 0.8000 g/cm³ Flash Pt: 11 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005913 CAS Number: 77-09-8 UN: 1993 ADR: 3,II |
Specification Test Results
Appearance | Clear, very light pink, Methanol odor |
Suitability for Use | Colorless (pH 8.0) - Pink (pH 10.0) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...