1,10-Phenanthroline 0.3% (w/v) in aqueous solution

Supplier: Ricca Chemical

R5522000-1A R5522000-500A
RC5522-32EA 161 USD
RC5522-32 RC5522-16
1,10-Phenanthroline 0.3% (w/v) in aqueous solution
Formula: C₁₂H₈N₂
MDL Number: MFCD00011678
CAS Number: 66-71-7
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5092a8ec81301ed6ed30f75429f9f27784db47a5&vid=106c4325bb7c310de902d3118cf4b31bcee3dc92&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR