Nitric acid 0.0200 N (N/50)

Supplier: Ricca Chemical

Synonyms: Nitric acid solution

R5385000-500A R5385000-1A
RC5385-16EA 35 USD
RC5385-16 RC5385-32
Nitric acid 0.0200 N (N/50)
Nitric acid
Formula: HNO₃
MW: 63.01
Boiling Pt: 100 °C
Melting Pt: 0.0 °C
Density: 1.51
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
Merck Index: 13,06610

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 84) 0.02000 ± 0.00002 N at 20.0°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR