Neocuproine 0.1% (w/v) in methanol, Laboratory Reagent

Supplier: Ricca Chemical
Danger

Synonyms: 2,9-Dimethyl-1,10-phenanthroline

R5240000-500A R5240000-120A
RC524016EA 149 USD
RC524016 RC5240-4
Neocuproine 0.1% (w/v) in methanol, Laboratory Reagent
Neocuproine
Formula: C₁₄H₁₂N₂
MW: 208.26 g/mol
Boiling Pt: ~ 64.5 °C
Melting Pt: –98 °C
Density: 0.8000 g/cm³
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00004973
CAS Number: 484-11-7
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear, colorless, Methanol odor

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR