Methyl red-methylene blue solution mixed indicator

Supplier: Ricca Chemical
Danger

R5164000-500A R5164000-120A
RC5164-16EA 85 USD
RC5164-16 RC5164-4
Methyl red-methylene blue solution mixed indicator
Methyl red-methylene blue
Boiling Pt: ~ 77 °C
Density: 0.8000 g/cm³
Storage Temperature: Ambient
UN: 1987
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear, dark green, alcohol odor
Suitability for Use Purple (pH 6.2)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR