Methyl red solution R
Supplier: Ricca Chemical
|
Danger
|
R5047000-120C
R5047000-500C
RC5047-4EA
61
USD
RC5047-4
RC5047-16
Methyl red solution R
Methyl red solution R
Formula:
C₁₅H₁₅N₃O₂ MW: 269.30 g/mol Boiling Pt: ~ 82 °C Density: 0.9138 g/cm³ Flash Pt: 24 °C Storage Temperature: Ambient |
UN:
1987 ADR: 3,III |
Specification Test Results
Appearance | Clear, dark red, alcoholic odor |
Suitability for Use | Red (pH 4.4) - Yellow (pH 6.0) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...