Methyl orange-xylene cyanol solution mixed indicator

Supplier: Ricca Chemical

R5030000-120C R5030000-500C
RC5030-4EA 70 USD
RC5030-4 RC5030-16
Methyl orange-xylene cyanol solution mixed indicator
Methyl orange-xylene cyanol

Methyl orange indicator solution, modified (methyl orange-xylene cyanol mixed indicator).

Formula: C₃₉H₄₁N₅Na₂O₁₀S₃
Boiling Pt: 100 °C
Melting Pt: 0.0 °C
Density: 1.0000 g/cm³
Storage Temperature: Ambient
CAS Number: 62758-15-0

Order Now

Specification Test Results

Appearance Clear, dark green, odorless
Suitability for Use Green (pH > 4.6) - Gray (pH 3.8) - Mauve (pH < 2.9)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR