4-(Methylamino)phenol sulfate USP reference solution (RS)
Supplier: Ricca Chemical
Synonyms:
p-(Methylamino)phenol sulphate, Metol, 4-Methylaminophenol sulphate, 4-(Methylamino)phenol hemisulphate, 4-Methylaminophenolsulphate, 4-(Methylamino)phenol hemisulphate salt
R4839000-120C
R4839000-500C
RC4839-4EA
67
USD
RC4839-4
RC4839-16
4-(Methylamino)phenol sulfate USP reference solution (RS)
4-(Methylamino)phenol sulphate
Formula:
C₁₄H₂₀N₂O₆S MW: 344.39 g/mol |
MDL Number:
MFCD00013140 CAS Number: 55-55-0 UN: 2811 ADR: 6.1,III |
Specification Test Results
Appearance | Clear, pale green, Bisulfite odor |
Suitability for Use | Blue in the presence of Phosphate |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...