Mercuric sulfate solution (red mercuric oxide) 80 g/L in 6 N Sulfuric Acid
Supplier: Ricca Chemical
|
Danger
|
R4780000-1A
R4780000-500A
RC4780-32EA
620
USD
RC4780-32
RC4780-16
Mercuric sulfate solution (red mercuric oxide) 80 g/L in 6 N Sulfuric Acid
Mercuric sulfate solution (red mercuric oxide)
Density:
1.2000 g/cm³ Storage Temperature: Ambient |
UN:
2922 ADR: 8 (6.1),II |
Specification Test Results
Appearance | Clear, colorless, odorless |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...