Mercury (II) potassium iodide (Mayer) USP Test Solution (TS)

Supplier: Ricca Chemical
Danger

R4765000-500B R4765000-1B
RC4765-16EA 94 USD
RC4765-16 RC4765-32
Mercury (II) potassium iodide (Mayer) USP Test Solution (TS)
Mercury (II) potassium iodide

Mercuric-potassium iodide Mayer's reagent.

Formula: HgI₄K₂
Boiling Pt: ~ 101 °C
Melting Pt: ~ 0 °C
Density: 1.0500 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00049656
CAS Number: 7783-33-7
UN: 3287
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Suitability for Use Yellow precipitate with Choline Chloride


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR