Manganese(II) sulphate

Supplier: Ricca Chemical

Synonyms: Manganese sulphate, Manganous sulphate

R4615000-500A R4615000-1A
RC4615-16EA 63 USD
RC4615-16 RC4615-32
Manganese(II) sulphate
Manganese(II) sulphate
Formula: MnO₄S
MW: 151
Density: 3.25
MDL Number: MFCD00003464
CAS Number: 7785-87-7
UN: 3077
ADR: 9 III
Merck Index: 12,05782

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 915) 0.00300 ± 0.00005 M at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR