Iodine monobromide USP Test Solution (TS)

Supplier: Ricca Chemical

R4102000-500C R4102000-120C
RC4102-16EA 319 USD
RC4102-16 RC4102-4
Iodine monobromide USP Test Solution (TS)
Iodine monobromide
Formula: IBr
MW: 206.81 g/mol
MDL Number: MFCD00011353
CAS Number: 7789-33-5
UN: 3260
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear, dark brown, Iodine/Bromine/Acetic odor
Total Halogen Assay at 20 °C (traceable to NIST SRM 136) 0.193 ± 0.002 N at 20.0 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR