8-Quinolinol 2%

Supplier: Ricca Chemical
Danger

Synonyms: 8-Hydroxyquinoline

R3940000-120C R3940000-500C
RC3940-4EA 30 USD
RC3940-4 RC3940-16
8-Quinolinol 2%
8-Quinolinol
Formula: C₉H₇NO
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0300 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00006807
CAS Number: 148-24-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=752471e0b5caa5507eb8e74e8054dc401912b3ac&vid=ff81328b844178e10beffb1867c97da0d03eda22&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance clear, yellow, odorless


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR