Formalin-aceto-alcohol(F-A-A) mixture for rapid fixation of tissue

Supplier: Ricca Chemical
Danger

R3210000-500A R3210000-1A
RC321016EA 81 USD
RC321016 RC3210-32
Formalin-aceto-alcohol(F-A-A) mixture for rapid fixation of tissue
Formalin-aceto-alcohol(F-A-A) mixture
Boiling Pt: ~ 82 °C
Density: 0.9200 g/cm³
Storage Temperature: Ambient
UN: 1987
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear, colorless, Formaldehyde odor


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR