Diphenylamine 0.3% (w/v) in aqueous solution
Supplier: Ricca Chemical
Synonyms:
N-Phenylaniline, N-Phenylbenzeneamine
R2585000-500C
R2585000-120C
RC2585-16EA
117
USD
RC2585-16
RC2585-4
Diphenylamine 0.3% (w/v) in aqueous solution
Diphenylamine
Formula:
(C₆H₅)₂NH MW: 169.23 g/mol |
MDL Number:
MFCD00003014 CAS Number: 122-39-4 UN: 3077 ADR: 9,III |
Specification Test Results
Appearance | Clear, colorless, odorless |
Suitability for Use | Colorless (reduced) - Red-violet (oxidized) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...