2',7'-Dichlorofluorescein 0.1% (w/v) in dilute alcohol USP Test Solution (TS)

Supplier: Ricca Chemical
Danger

Synonyms: 2',7'-Dichloro-3',6'-dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one

R2549000-120A R2549000-500A
RC2549-4EA 93 USD
RC2549-4 RC2549-16
2',7'-Dichlorofluorescein 0.1% (w/v) in dilute alcohol USP Test Solution (TS)
2',7'-Dichlorofluorescein
Formula: C₂₀H₁₀Cl₂O₅
MW: 401.20 g/mol
Boiling Pt: ~ 82 °C
Density: 0.9000 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00005047
CAS Number: 76-54-0
UN: 1987
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear, greenish yellow liquid
Suitability for Use Adsorption: greenish-yellow (excess Chloride) - Rose (excess Silver ions)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR