Cupric sulfate 12.5% (w/v) in aqueous solution USP Test Solution (TS)
Supplier: Ricca Chemical
|
Danger
|
R2494000-120A
R2494000-500A
RC2494-4EA
49
USD
RC2494-4
RC2494-16
Cupric sulfate 12.5% (w/v) in aqueous solution USP Test Solution (TS)
Copper(II) sulphate solutions
Formula:
CuSO₄·5H₂O Boiling Pt: ~ 100 °C Melting Pt: ~ 0 °C Density: 1.0500 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00149681 CAS Number: 7758-99-8 |
Specification Test Results
Appearance | Clear, blue, odorless |
Assay at 20 °C (traceable to NIST SRM 136) | 125.0 ± 1.0 g/L |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...