Cupric citrate USP Test Solution (TS), alkaline

Supplier: Ricca Chemical
Danger

R2493000-120A R2493000-500A
RC2493-4EA 186 USD
RC2493-4 RC2493-16
Cupric citrate USP Test Solution (TS), alkaline
Cupric citrate
Formula: C₆H₄Cu₂O₇
Boiling Pt: ~ 100 °C
Density: 1.1000 g/cm³
Storage Temperature: Ambient
CAS Number: 866-82-0

Order Now

Specification Test Results

Appearance Clear, blue, odorless
Suitability for Use Suitable for reducing sugar determination


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR