Color reagent for nitrate determination

Supplier: Ricca Chemical
Danger

R2233000-500C R2233000-120C
RC2233-16EA 146 USD
RC2233-16 RC2233-4
Color reagent for nitrate determination
Color reagent
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0500 g/cm³
Storage Temperature: Refrigerator
UN: 3264
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear, tan-brown, odorless
Suitability for Use Red color with Nitrite


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR