Carmine reagent for determination of boron

Supplier: Ricca Chemical
Danger

Synonyms: Carminic acid, Cochineal, Alum lake of Carminic Acid, Natural Red 4, FD and C Carmine, CI 75470, Carmine, calcium/aluminum lake

R1850000-120C R1850000-500C1 R1850000-1C1
RC1850-4EA 65 USD
RC1850-4 RC1850-16 RC1850-32
Carmine reagent for determination of boron
Carmine

Carmine Reagent, for Boron determination


  • APHA Standard Methods Reagents
  • Shelf Life 12 months

Formula: C₂₂H₂₀O₁₃
MW: 492.389 g/mol
Boiling Pt: ~ 338 °C
Density: 1.8000 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00167028
CAS Number: 1390-65-4
UN: 1830
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear, red, odorless

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR