Bromocresol green-methyl red USP Test Solution (TS)

Supplier: Ricca Chemical
Danger

Synonyms: Methyl Red - Bromocresol green solution

R1221000-120A R1221000-500A
RC1221-4EA 92 USD
RC1221-4 RC1221-16
Bromocresol green-methyl red USP Test Solution (TS)
Bromocresol green-methyl red
Boiling Pt: ~ 77 °C
Density: 0.8300 g/cm³
Flash Pt: ~ 69°F
Storage Temperature: Ambient
MDL Number: MFCD00134126
UN: 1987
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear, red, alcohol odor
Suitability for Use Greenish-Blue (pH 5.2) - Light Pink (pH 4.5), with Gray warning color

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR