Bromate-Bromide solution 0.0250 N (N/40)

Supplier: Ricca Chemical
Warning

R1166000-4C R1166000-1C
RC1166-1EA 306.64 USD
RC1166-1 RC1166-32
Bromate-Bromide solution 0.0250 N (N/40)
Bromate-Bromide

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 136) 0.02500 ± 0.00002 N at 20.0 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR